Ethyl-2-oxo-3-piperidincarboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-Piperidone-3-carboxylate, 2-Oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 2-Oxo-3-piperidinecarboxylate, 3-Ethoxycarbonyl-2-piperidone

BIB6023-100G BIB6023-250G BIB6023-25G
APOSBIB6023-100GEA 400 EUR
APOSBIB6023-100G APOSBIB6023-250G APOSBIB6023-25G
Ethyl-2-oxo-3-piperidincarboxylat
3-Carbethoxy-2-piperidone
Formel: C8H13NO3
Molecular Weight: 171,1900 g/mol
Schmelzpunkt: 80…82 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00006038
CAS-Nummer: 3731-16-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR