4-Chloroindole-3-acetic acid

Lieferant: Apollo Scientific
Achtung

BIB6028-250MG BIB6028-500MG
APOSBIB6028-250MGEA 40.6 EUR
APOSBIB6028-250MG APOSBIB6028-500MG
4-Chloroindole-3-acetic acid
4-Chloroindole-3-acetic acid
Formel: C₁₀H₈ClNO₂
Molecular Weight: 209.63 g/mol
Schmelzpunkt: 179…188 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00216155
CAS-Nummer: 2519-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR