1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine
Lieferant: Apollo Scientific
Synonyme: 1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine, 1,2-Dihexadecanoyl-sn-glycero-3-phosphoethanolamine, Dipalmitoylphosphatidylethanolamine, 2-Aminoethyl (R)-2,3-Bis(hexadecanoyloxy)propyl Hydrogen Phosphate, (2R)-3-(((2-Aminoethoxy)(hydroxy)phosphoryl)oxy)propane-1,2-diyl dipalmitate, L-DPPE
BIB6045-100MG BIB6045-250MG BIB6045-500MG
APOSBIB6045-100MGEA 161 EUR
APOSBIB6045-100MG APOSBIB6045-250MG APOSBIB6045-500MG
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine
1,2-Dipalmitoyl-sn-glycero-3-phosphoethanolamine
Formel: C₃₇H₇₄NO₈P
Molecular Weight: 691,97 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036776
CAS-Nummer: 923-61-5