meso-Erythritol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
meso-Erythritol, (2R,3S)-rel-Butane-1,2,3,4-tetraol, i-Erythritol
BIB6050-1KG
BIB6050-500G
APOSBIB6050-1KGEA
75.4
EUR
APOSBIB6050-1KG
APOSBIB6050-500G
meso-Erythritol
meso-Erythritol
Formel:
C₄H₁₀O₄ Molecular Weight: 122,12 g/mol Siedepunkt: 329…331 °C Schmelzpunkt: 120…123 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00004710 CAS-Nummer: 149-32-6 EINECS: 205-737-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...