3-Indoxyl phosphate disodium salt

Lieferant: Apollo Scientific

Synonyme: 3-Indolyl phosphate disodium salt

BIB6081-1G BIB6081-500MG BIB6081-5G
APOSBIB6081-1GEA 313 EUR
APOSBIB6081-1G APOSBIB6081-500MG APOSBIB6081-5G
3-Indoxyl phosphate disodium salt
3-Indoxyl phosphate disodium salt
Formel: C₈H₆NNa₂O₄P
Molecular Weight: 257.09 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00040646
CAS-Nummer: 3318-43-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR