L-Kynurenine freie Base

Lieferant: Apollo Scientific
Achtung

Synonyme: β-Anthraniloyl-L-alanine, (S)-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, L-2-Amino-4-(2-aminophenyl)-4-oxobutanoic acid, Kynurenine

BIB6088-1G BIB6088-100MG
APOSBIB6088-1GEA 574 EUR
APOSBIB6088-1G APOSBIB6088-100MG
L-Kynurenine freie Base
L-Kynurenine
Formel: C₁₀H₁₂N₂O₃
Molecular Weight: 208,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00069912
CAS-Nummer: 2922-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR