D-(+)-Melezitose monohydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,2'R,3S,3'S,4S,4'S,5R,5'R,6R,6'R)-6,6'-(((2S,3S,4R,5R)-4-Hydroxy-2,5-bis(hydroxymethyl)tetrahydrofuran-2,3-diyl)bis(oxy))bis(2-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol) hydrate

BIB6097-100G BIB6097-250G
APOSBIB6097-100GEA 574 EUR
APOSBIB6097-100G APOSBIB6097-250G
D-(+)-Melezitose monohydrate
D-(+)-Melezitose monohydrate
Formel: C₁₈H₃₄O₁₇
Molecular Weight: 522.453 g/mol
Schmelzpunkt: 156 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00149448
CAS-Nummer: 10030-67-8
EINECS: 209-894-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR