2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-MUNANA

BIB6114-100MG BIB6114-25MG
APOSBIB6114-100MGEA 393 EUR
APOSBIB6114-100MG APOSBIB6114-25MG
2′-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic acid sodium salt hydrate
2′-(4-Methylumbelliferyl)-alpha-D-N-acetylneuraminic acid sodium salt hydrate
Formel: C₂₁H₂₄NNaO₁₁·xH₂O
Molecular Weight: 489,41 g/mol (anhy)
Lagertemperatur: Gefrierschrank
MDL: MFCD00057309
CAS-Nummer: 76204-02-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR