DL-Glyceraldehyde dimer 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,6-Dihydroxy-1,4-dioxane-2,5-dimethanol, (2R,3S,5S,6R)-3,6-Bis(hydroxymethyl)-1,4-dioxane-2,5-diol

BIB6213-500MG BIB6213-1G BIB6213-5G
APOSBIB6213-500MGEA 43.5 EUR
APOSBIB6213-500MG APOSBIB6213-1G APOSBIB6213-5G
DL-Glyceraldehyde dimer 95+%
DL-Glyceraldehyde dimer
Formel: C₆H₁₂O₆
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 144…145 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00070528
CAS-Nummer: 23147-59-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR