Sodium phenylpyruvate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Oxo-3-phenylpropionic acid sodium salt, Phenylpyruvic acid sodium salt

BIB6259-100G BIB6259-50G BIB6259-25G
APOSBIB6259-100GEA 502 EUR
APOSBIB6259-100G APOSBIB6259-50G APOSBIB6259-25G
Sodium phenylpyruvate
Sodium phenylpyruvate
Formel: C₉H₇NaO₃
Molecular Weight: 186,14 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00002590
CAS-Nummer: 114-76-1
EINECS: 204-053-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR