Pyr-βNA
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(S)-N-(Naphthalen-2-yl)-5-oxopyrrolidine-2-carboxamide, L-Pyroglutamic acid beta-naphthylamide
BIB6266-5G
BIB6266-1G
BIB6266-250MG
APOSBIB6266-5GEA
729
EUR
APOSBIB6266-5G
APOSBIB6266-1G
APOSBIB6266-250MG
Pyr-βNA
Pyr-βNA
Formel:
C₁₅H₁₄N₂O₂ Molecular Weight: 254.28 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00055911 CAS-Nummer: 22155-91-5 EINECS: 244-809-9 UN: 3077 ADR: 9+EHS III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...