Atractyloside potassium salt

Lieferant: Apollo Scientific
Gefahr

Synonyme: Potassium (2R,4aR,6aR,7S,9R,11aS,11bR)-7-hydroxy-2-(((2R,3R,4R,5R,6R)-6-(hydroxymethyl)-3-((3-methylbutanoyl)oxy)-5-(sulphonatooxy)-4-(sulphooxy)tetrahydro-2H-pyran-2-yl)oxy)-11b-methyl-8-methylenetetradecahydro-6a,9-methanocyclohepta[a]naphthalene-4-carbox, (2β)-15α-hydroxy-2-[[2-O-(3-methyl-1-oxobutyl)-3,4-di-O-sulpho-β-D-glucopyranosyl]oxy]-19-norkaur-16-en-18-oic acid, dipotassium salt

BIB6292-100MG BIB6292-25MG BIB6292-50MG
APOSBIB6292-100MGEA 641 EUR
APOSBIB6292-100MG APOSBIB6292-25MG APOSBIB6292-50MG
Atractyloside potassium salt
Atractyloside potassium salt
Formel: C₃₀H₄₄K₂O₁₆S₂
Molecular Weight: 802,99 g/mol
Schmelzpunkt: 174 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00078810
CAS-Nummer: 102130-43-8
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR