Cyanidin-3-O-glucoside chloride
Lieferant: Apollo Scientific
Synonyme:
Kuromanine Chloride, 2-(3,4-Dihydroxyphenyl)-3-(β-D-glucopyranosyloxy)-5,7-dihydroxy-1-benzopyrylium Chloride
BIB6297-10MG
BIB6297-5MG
APOSBIB6297-10MGEA
284
EUR
APOSBIB6297-10MG
APOSBIB6297-5MG
Cyanidin-3-O-glucoside chloride
Cyanidin-3-O-glucoside chloride
Formel:
C₂₁H₂₁O₁₁Cl Molecular Weight: 484.84 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00064923 CAS-Nummer: 7084-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...