Deferoxamine mesylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
DFOM
BIB6311-10G
BIB6311-5G
BIB6311-1G
APOSBIB6311-10GEA
348
EUR
APOSBIB6311-10G
APOSBIB6311-5G
APOSBIB6311-1G
Deferoxamine mesylate
Deferoxamine mesylate
Formel:
C₂₆H₅₂N₆O₁₁S Molecular Weight: 656,793 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00058605 CAS-Nummer: 138-14-7 EINECS: 205-314-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...