Fmoc-D-isoleucine
Lieferant: Apollo Scientific
Synonyme:
Fmoc-D-Ile-OH, (2R,3R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-methylpentanoic acid
BIBA1019-1G
BIBA1019-250MG
BIBA1019-5G
APOSBIBA1019-1GEA
306
EUR
APOSBIBA1019-1G
APOSBIBA1019-250MG
APOSBIBA1019-5G
Fmoc-D-isoleucine
Fmoc-D-isoleucine
Formel:
C₂₁H₂₃NO₄ Molecular Weight: 353,418 g/mol Schmelzpunkt: 143…153 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00080291 CAS-Nummer: 143688-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...