H-L-Dap(boc)-OMe·HCl

Lieferant: Apollo Scientific

Synonyme: H-Dapa(Boc)-OMe.HCl, H-Dap(Boc)-OMe·HCl

BIBA1021-1G BIBA1021-5G BIBA1021-10G
APOSBIBA1021-1GEA 102 EUR
APOSBIBA1021-1G APOSBIBA1021-5G APOSBIBA1021-10G
H-L-Dap(boc)-OMe·HCl
H-L-Dap(boc)-OMe·HCl
Formel: C₉H₁₉ClN₂O₄
Molecular Weight: 254,711 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00798684
CAS-Nummer: 114559-25-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR