L-4-Benzoylphenylalanine
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-3-(4-benzoylphenyl)propanoic acid, H-Bpa-OH, H-p-Bz-Phe-OH, H-Phe(4-Bz)-OH
BIBA1027-1G
BIBA1027-5G
BIBA1027-10G
APOSBIBA1027-1GEA
139
EUR
APOSBIBA1027-1G
APOSBIBA1027-5G
APOSBIBA1027-10G
L-4-Benzoylphenylalanine
L-4-Benzoylphenylalanine
Formel:
C₁₆H₁₅NO₃ Schmelzpunkt: 180 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00238102 CAS-Nummer: 104504-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...