Fmoc-3-(aminooxy)acetic acid ≥98%

Lieferant: Apollo Scientific

Synonyme: Fmoc-Aoa, Fmoc-Aoa-OH

BIBP1076-500MG BIBP1076-1G
APOSBIBP1076-500MGEA 66.7 EUR
APOSBIBP1076-500MG APOSBIBP1076-1G
Fmoc-3-(aminooxy)acetic acid ≥98%
Fmoc-3-(aminooxy)acetic acid
Formel: C₁₇H₁₅NO₅
Molecular Weight: 313,31 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01076133
CAS-Nummer: 123106-21-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR