Bafilomycin B1

Lieferant: Apollo Scientific

BIBR1013-1MG BIBR1013-5MG
APOSBIBR1013-1MGEA 236 EUR
APOSBIBR1013-1MG APOSBIBR1013-5MG
Bafilomycin B1
Bafilomycin B1
Formel: C₄₄H₆₅NO₁₃
Molecular Weight: 815,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00242671
CAS-Nummer: 88899-56-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR