Enniatin B1

Lieferant: Apollo Scientific

BIBR1041-1MG BIBR1041-5MG
APOSBIBR1041-1MGEA 348 EUR
APOSBIBR1041-1MG APOSBIBR1041-5MG
Enniatin B1
Enniatin B1
Formel: C₃₄H₅₉N₃O₉
Lagertemperatur: Raumtemperatur
CAS-Nummer: 19914-20-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR