Piericidin A

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-((2E,5E,7E,9R,10R,11E)-10-Hydroxy-3,7,9,11-tetramethyltrideca-2,5,7,11-tetraen-1-yl)-5,6-dimethoxy-3-methylpyridin-4-ol, SN 198E, Piericidin A ethanol solution, IT 143D, Shaoguanmycin B, Piericidin A1, MT 1882-I

BIBR1077-1MG BIBR1077-5MG
APOSBIBR1077-1MGEA 203 EUR
APOSBIBR1077-1MG APOSBIBR1077-5MG
Piericidin A
Piericidin A
Formel: C₂₅H₃₇NO₄
Molecular Weight: 415,6 g/mol
Flammpunkt: 85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01677956
CAS-Nummer: 2738-64-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR