Cefazolin sodium salt
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Sodium (6R,7R)-7-(2-(1H-tetrazol-1-yl)acetamido)-3-(((5-methyl-1,3,4-thiadiazol-2-yl)thio)methyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate
BIC0101-1G
BIC0101-5G
APOSBIC0101-1GEA
52.2
EUR
APOSBIC0101-1G
APOSBIC0101-5G
Cefazolin sodium salt
Cefazolin sodium salt
Formel:
C₁₄H₁₄N₈NaO₄S₃ Molecular Weight: 476.48 g/mol Schmelzpunkt: 170…172 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00056883 CAS-Nummer: 27164-46-1 EINECS: 248-278-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...