Cefotaxime sodium salt

Lieferant: Apollo Scientific
Gefahr

Synonyme: Sodium (6R,7R)-3-(acetoxymethyl)-7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cefotaxime sodium

BIC0111-25G BIC0111-5G BIC0111-100G
APOSBIC0111-25GEA 132 EUR
APOSBIC0111-25G APOSBIC0111-5G APOSBIC0111-100G
Cefotaxime sodium salt
Cefotaxime sodium salt

Cefotaxime is an inhibitor of bacterial cell wall synthesis.

Formel: C₁₆H₁₇N₅O₇S₂
Molecular Weight: 477,44 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00079073
CAS-Nummer: 64485-93-4
EINECS: 264-915-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR