Cephalothin sodium salt

Lieferant: Apollo Scientific
Gefahr

Synonyme: Sodium (6R,7R)-3-(acetoxymethyl)-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate, Cephalothin sodium

BIC0115-10G BIC0115-25G BIC0115-50G
APOSBIC0115-10GEA 319 EUR
APOSBIC0115-10G APOSBIC0115-25G APOSBIC0115-50G
Cephalothin sodium salt
Cephalothin sodium salt

First generation cephalosporin antibiotic. Disrupts the synthesis of the peptidoglycan layer of bacterial cell walls. It is effective against gram-positive and gram-negative bacteria.

Formel: C₁₆H₁₅N₂NaO₆S₂
Molecular Weight: 418,414 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00072025
CAS-Nummer: 58-71-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR