L(-)-Cystin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(2R,2'R)-(-)-3,3'-Dithiobis(2-aminopropansäure)
, (R)-(-)-Cystin
, (H-Cys-OH)2
APOSBIC0670-1KGEA
82.9
EUR
APOSBIC0670-1KG
APOSBIC0670-100G
L(-)-Cystin
L(-)-Cystin
Formel:
HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH Molecular Weight: 240,3 g/mol Schmelzpunkt: >240 °C Dichte: 1,68 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064228 CAS-Nummer: 56-89-3 EINECS: 200-296-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...