L(+)-Cystein Hydrochlorid
Lieferant: Apollo Scientific
Achtung
Synonyme: H-Cys-OH.HCl, (R)-(+)-Cystein Hydrochlorid, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid
BIC0707-1KG BIC0707-500G
APOSBIC0707-1KGEA 92.8 EUR
APOSBIC0707-1KG APOSBIC0707-500G
L(+)-Cystein Hydrochlorid
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Molecular Weight: 157,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
EINECS: 200-157-7