L(+)-Cystein
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(R)-(+)-2-Amino-3-mercaptopropansäure, H-Cys-OH, (R)-(+)-Cystein
BIC0709-100G
BIC0709-1KG
BIC0709-250G
APOSBIC0709-100GEA
21.9
EUR
APOSBIC0709-100G
APOSBIC0709-1KG
APOSBIC0709-250G
L(+)-Cystein
L(+)-Cystein
Formel:
HSCH₂CH(NH₂)CO₂H Molecular Weight: 121,16 g/mol Schmelzpunkt: 240 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064306 CAS-Nummer: 52-90-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...