Chelerythrine chloride
Lieferant: Apollo Scientific
Synonyme:
1,2-Dimethoxy-12-methyl-[1,3]dioxolo[4',5':4,5]benzo[1,2-c]phenanthridin-12-ium chloride
BIC1008-50MG
BIC1008-100MG
BIC1008-500MG
APOSBIC1008-50MGEA
109
EUR
APOSBIC1008-50MG
APOSBIC1008-100MG
APOSBIC1008-500MG
Chelerythrine chloride
Chelerythrine chloride
Formel:
C₂₁H₁₈ClNO₄ Molecular Weight: 383,828 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00060717 CAS-Nummer: 3895-92-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...