Cyclopamine

Lieferant: Apollo Scientific
Achtung

APOSBIC1011-250MGEA 212 EUR
APOSBIC1011-250MG APOSBIC1011-500MG APOSBIC1011-25MG APOSBIC1011-100MG APOSBIC1011-10MG APOSBIC1011-5MG
Cyclopamine
Cyclopamine
Formel: C₂₇H₄₁NO₂
Molecular Weight: 411,63 g/mol
Schmelzpunkt: 225…230 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD09878269
CAS-Nummer: 4449-51-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR