Cytochalasin A
Lieferant: Apollo Scientific
|
Gefahr
|
BIC1013-10MG
BIC1013-1MG
BIC1013-5MG
APOSBIC1013-10MGEA
725
EUR
APOSBIC1013-10MG
APOSBIC1013-1MG
APOSBIC1013-5MG
Cytochalasin A
Cytochalasin A
Oxidised analog of cytochalasin B (BIC1014).
Formel:
C₂₉H₃₅NO₅ Molecular Weight: 477,6 g/mol Schmelzpunkt: 193…195 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00005935 CAS-Nummer: 14110-64-6 UN: 1544 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...