5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Dioxopyrrolidin-1-yl 3',6'-dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-5-carboxylate, 5-FAM-N-succinimidyl ester, 5-Carboxyfluorescein NHS ester, 5-FAM SE

BIC1054-100MG
APOSBIC1054-100MGEA 200 EUR
APOSBIC1054-100MG
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE) 98%
5-Carboxyfluorescein-N-succinimidyl ester (5-FAM SE)
Formel: C₂₅H₁₅NO₉
Molecular Weight: 473,39 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00077322
CAS-Nummer: 92557-80-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR