5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]
Lieferant: Apollo Scientific
Achtung
Synonyme: CFDA-SE, CFSE, Carboxyfluorescein diacetate succinimidyl ester, 5(6)-CFDA N-succinmidyl ester, 5(6)-Carboxyfluorescein diacetate succinimidyl ester, 5(6)-CFDA SE
BIC1063-100MG BIC1063-50MG
APOSBIC1063-100MGEA 386 EUR
APOSBIC1063-100MG APOSBIC1063-50MG
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]
5(6)-Carboxyfluorescein diacetate NHS ester [CFSE (5(6)-CFDA SE)]
Formel: C₂₉H₁₉NO₁₁
Molecular Weight: 557.467 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00037466
CAS-Nummer: 150347-59-4