5(6)-Carboxyfluorescein diacetate

Lieferant: Apollo Scientific

Synonyme: CFDA, 5(6)-Carboxy-di-O-acetylfluorescein, 5(6)-CFDA

BIC1066-100MG BIC1066-250MG
APOSBIC1066-100MGEA 284 EUR
APOSBIC1066-100MG APOSBIC1066-250MG
5(6)-Carboxyfluorescein diacetate
5(6)-Carboxyfluorescein diacetate
Formel: C₂₅H₁₆O₉
Molecular Weight: 460.39 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00467494
CAS-Nummer: 124387-19-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR