Cytidin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

BIC2010-10G BIC2010-250G BIC2010-50G
APOSBIC2010-10GEA 21.9 EUR
APOSBIC2010-10G APOSBIC2010-250G APOSBIC2010-50G
Cytidin
Cytidin
Formel: C₉H₁₃N₃O₅
Molecular Weight: 243,22 g/mol
Schmelzpunkt: 210…220 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006545
CAS-Nummer: 65-46-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR