Coenzyme A trilithium salt

Lieferant: Apollo Scientific
Achtung

BIC7401-100MG BIC7401-500MG BIC7401-250MG
APOSBIC7401-100MGEA 179 EUR
APOSBIC7401-100MG APOSBIC7401-500MG APOSBIC7401-250MG
Coenzyme A trilithium salt
Coenzyme A trilithium salt
Formel: C₂₁H₃₃N₇O₁₆P₃SLi₃
Lagertemperatur: Gefrierschrank
MDL: MFCD00075848
CAS-Nummer: 18439-24-2
EINECS: 242-317-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR