SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
Lieferant: Apollo Scientific
Achtung
Synonyme: SMCC
BICL206-100MG BICL206-25MG
APOSBICL206-100MGEA 94.4 EUR
APOSBICL206-100MG APOSBICL206-25MG
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
SMCC (N-Succinimidyl 4-(N-Maleimidomethyl)cyclohexanecarboxylate)
Primary amine and Sulphhydryl reactive, Enzyme antibody conjugation.
Formel: C₁₆H₁₈N₂O₆
Molecular Weight: 334,33 g/mol
Schmelzpunkt: 180…182 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00009634
CAS-Nummer: 64987-85-5