N-Butyldeoxynojirimycin 99% min

Lieferant: Apollo Scientific

Synonyme: (2R,3R,4R,5S)-1-Butyl-2-(hydroxymethyl)piperidine-3,4,5-triol, N-(n-Butyl)-1-deoxynojirimycin, Miglustat

BICL4040-10MG BICL4040-25MG BICL4040-5MG
APOSBICL4040-10MGEA 420 EUR
APOSBICL4040-10MG APOSBICL4040-25MG APOSBICL4040-5MG
N-Butyldeoxynojirimycin 99% min
N-Butyldeoxynojirimycin
Formel: C₁₀H₂₁NO₄
Molecular Weight: 219.28 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00272581
CAS-Nummer: 72599-27-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR