2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-lactosyl bromide 98% min

Lieferant: Apollo Scientific
Achtung

Synonyme: (2R,3S,4S,5R,6S)-2-(Acetoxymethyl)-6-(((2R,3R,4S,5R,6R)-4,5-diacetoxy-2-(acetoxymethyl)-6-bromotetrahydro-2H-pyran-3-yl)oxy)tetrahydro-2H-pyran-3,4,5-triyl triacetate, 2,2',3,3',4',6,6'-HEPTA-O-ACETYL-α-D-LACTOSYL BROMIDE, 2,3,6,2 ,3 ,4 ,6 -Hepta-O-acetyl-a-D-cellobiosyl bromide, Bromo Heptaacetyl-D-lactoside, Acetobromo-α-D-lactoside

BICL4047-1G BICL4047-5G BICL4047-2G
APOSBICL4047-1GEA 212 EUR
APOSBICL4047-1G APOSBICL4047-5G APOSBICL4047-2G
2,3,6,2',3',4',6'-Hepta-O-acetyl-α-D-lactosyl bromide 98% min
2,3,6,2',3',4',6'-Hepta-O-acetyl-alpha-D-lactosyl bromide
Formel: C₂₆H₃₅BrO₁₇
Molecular Weight: 699,45 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00153593
CAS-Nummer: 4753-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR