2-NBDG 95% min

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-​Deoxy-​2-​[(7-​nitro-​2,​1,​3-​benzoxadiazol-​4-​yl)amino]-​D-​glucose, NBD-Glucose, (2R,3R,4S,5R)-3,4,5,6-Tetrahydroxy-2-((7-nitrobenzo[c][1,2,5]oxadiazol-4-yl)amino)hexanal

BICL4185-10MG BICL4185-5MG
APOSBICL4185-10MGEA 522 EUR
APOSBICL4185-10MG APOSBICL4185-5MG
2-NBDG 95% min
2-NBDG
Formel: C₁₂H₁₄N₄O₈
Molecular Weight: 342,3 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD23135882
CAS-Nummer: 186689-07-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR