Cyanidin-3-O-galactoside chloride 95% min
Lieferant: Apollo Scientific
Synonyme:
2-(3,4-Dihydroxyphenyl)-5,7-dihydroxy-3-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)chromenylium chloride
BICL4237-5MG
BICL4237-1MG
APOSBICL4237-5MGEA
163
EUR
APOSBICL4237-5MG
APOSBICL4237-1MG
Cyanidin-3-O-galactoside chloride 95% min
Cyanidin-3-O-galactoside chloride
Formel:
C₂₁H₂₁O₁₁Cl Molecular Weight: 484.85 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00270387 CAS-Nummer: 27661-36-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...