Cholesterol-3-sulphate sodium salt 95% min

Lieferant: Apollo Scientific

Synonyme: Cholesteryl sodium sulphate, Cholesteryl sulphate sodium

BICL4238-1G BICL4238-250MG BICL4238-500MG
APOSBICL4238-1GEA 1200 EUR
APOSBICL4238-1G APOSBICL4238-250MG APOSBICL4238-500MG
Cholesterol-3-sulphate sodium salt 95% min
Cholesterol-3-sulphate sodium salt
Formel: C₂₇H₄₅O₄S·Na
Molecular Weight: 488,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00079091
CAS-Nummer: 2864-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR