1,2,4,6-Tetra-O-acetyl-3-O-allyl-β-D-glucopyranose 98% min

Lieferant: Apollo Scientific

BICL4369-1G BICL4369-5G BICL4369-2G
APOSBICL4369-1GEA 266 EUR
APOSBICL4369-1G APOSBICL4369-5G APOSBICL4369-2G
1,2,4,6-Tetra-O-acetyl-3-O-allyl-β-D-glucopyranose 98% min
1,2,4,6-Tetra-O-acetyl-3-O-allyl-beta-D-glucopyranose
Formel: C₁₇H₂₄O₁₀
Molecular Weight: 388,37 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 39698-00-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR