4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 92%
Lieferant: Apollo Scientific
Achtung
Synonyme: 2-(4-Amidinophenyl)indole-6-carboxamidine Dihydrochloride, 2-(4-Carbamimidoylphenyl)-1H-indole-6-carboximidamide dihydrochloride, DAPI.2HCl, 2-(4-Carbamimidoylphenyl)indole-6-carboximidamide Dihydrochloride, DAPI 2HCl
BID0433-100MG BID0433-250MG BID0433-1G
APOSBID0433-100MGEA 55.1 EUR
APOSBID0433-100MG APOSBID0433-250MG APOSBID0433-1G
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI) 92%
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)
Dye used in the staining of DNA.
Formel: C₁₆H₁₅N₅·2HCl
Molecular Weight: 350,25 g/mol
Schmelzpunkt: 330 °C (Zers.)
Lagertemperatur: Kühlschrank
MDL: MFCD00012681
CAS-Nummer: 28718-90-3
EINECS: 249-186-7