Deoxynivalenol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3α,7α,15-Trihydroxy-12,13-epoxytrichothec-9-en-8-one, DON, Vomitoxin

BID1001-1MG BID1001-5MG
APOSBID1001-1MGEA 203 EUR
APOSBID1001-1MG APOSBID1001-5MG
Deoxynivalenol
Deoxynivalenol
Formel: C₁₅H₂₀O₆
Molecular Weight: 296,3 g/mol
Schmelzpunkt: 147…153 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 51481-10-8
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR