Dihydrocytochalasin B

Lieferant: Apollo Scientific
Gefahr

BID1004-1MG BID1004-5MG
APOSBID1004-1MGEA 319 EUR
APOSBID1004-1MG APOSBID1004-5MG
Dihydrocytochalasin B
Dihydrocytochalasin B
Formel: C₂₉H₃₉NO₅
Molecular Weight: 481,63 g/mol
Schmelzpunkt: 203…205 °C
Lagertemperatur: Gefrierschrank
CAS-Nummer: 39156-67-7
EINECS: 254-324-4
UN: 2811
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR