2'-Desoxycytidin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one, deoxyC

BID1650-25G BID1650-50G BID1650-100G
APOSBID1650-25GEA 72.5 EUR
APOSBID1650-25G APOSBID1650-50G APOSBID1650-100G
2'-Desoxycytidin
2'-Desoxycytidin
Formel: C₉H₁₃N₃O₄
Molecular Weight: 227,22 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00006547
CAS-Nummer: 951-77-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR