Decyl-β-D-glucopyranosid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Decyl-β-D-glucosid
BID6001-1G
BID6001-5G
BID6001-10G
APOSBID6001-1GEA
95.7
EUR
APOSBID6001-1G
APOSBID6001-5G
APOSBID6001-10G
Decyl-β-D-glucopyranosid
Decyl-β-D-glucopyranosid
Formel:
C₁₆H₃₂O₆ Molecular Weight: 320,43 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063297 CAS-Nummer: 58846-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...