Diosmetin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5,7-Dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-4H-chromen-4-one, 5,7,3'-Trihydroxy-4'-methoxyflavone
BIDF1026-100MG
BIDF1026-250MG
APOSBIDF1026-100MGEA
79.9
EUR
APOSBIDF1026-100MG
APOSBIDF1026-250MG
Diosmetin
Diosmetin
Formel:
C₁₆H₁₂O₆ Schmelzpunkt: 259…264 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00017425 CAS-Nummer: 520-34-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...