Piperlongumine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(E)-1-(3-(3,4,5-Trimethoxyphenyl)acryloyl)-5,6-dihydropyridin-2(1H)-one
BIDF1035-250MG
BIDF1035-50MG
BIDF1035-100MG
APOSBIDF1035-250MGEA
174
EUR
APOSBIDF1035-250MG
APOSBIDF1035-50MG
APOSBIDF1035-100MG
Piperlongumine
Piperlongumine
Natural product.
Formel:
C₁₇H₁₉NO₅ Molecular Weight: 317,341 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00075706 CAS-Nummer: 20069-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...