Plumbagin

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Hydroxy-2-methylnaphthalene-1,4-dione, 5-Hydroxy-2-methyl-1,4-naphthoquinone

BIDF1037-1G
APOSBIDF1037-1GEA 299 EUR
APOSBIDF1037-1G
Plumbagin
Plumbagin

Natural product.

Formel: C₁₁H₈O₃
Molecular Weight: 188,18 g/mol
Schmelzpunkt: 76…78 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00001682
CAS-Nummer: 481-42-5
EINECS: 207-569-6
UN: 2923
ADR: 8+6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR